COMH04237AC6 Combi-Blocks, Inc.

N-([(3-Nitrophenyl)amino]carbonyl)glycine

  • MDL number MFCD08442839
  • Cas Number : 67513-14-8
  • Molecular Weight : 239.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2598
Availability details for
Quantity
  • Availability
  • Add to Cart