COMH04237AC7 Combi-Blocks, Inc.

1,3-Dimethyl-6-thien-2-yl-1h-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08558202
  • Cas Number : 893645-77-7
  • Molecular Weight : 273.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2599
Availability details for
Quantity
  • Availability
  • Add to Cart