COMH04237AED Combi-Blocks, Inc.

3-[(2-Methylbenzyl)oxy]benzoic acid

  • MDL number MFCD08443025
  • Cas Number : 926228-86-6
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2637
Availability details for
Quantity
  • Availability
  • Add to Cart