COMH04237AFC Combi-Blocks, Inc.

1-Bromo-3-(sulfinylamino)benzene

  • MDL number MFCD26792680
  • Cas Number : 89379-48-6
  • Molecular Weight : 218.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2652
Availability details for
Quantity
  • Availability
  • Add to Cart