COMH04237B13 Combi-Blocks, Inc.

[5-(3-Chloro-4-fluorophenyl)-2-furyl]methanol

  • MDL number MFCD05843218
  • Cas Number : 847482-06-8
  • Molecular Weight : 226.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2675
Availability details for
Quantity
  • Availability
  • Add to Cart