COMH04237B4D Combi-Blocks, Inc.

3-(3,4-Dimethyl-phenyl)-adamantane-1-carboxylic acid

  • MDL number MFCD01674000
  • Cas Number : 56531-57-8
  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2734
Availability details for
Quantity
  • Availability
  • Add to Cart