COMH04237B51 Combi-Blocks, Inc.

2-Methyl-3-([(4-methylphenyl)sulfonyl]amino)benzoic acid

  • MDL number MFCD06092540
  • Cas Number : 830349-15-0
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2738

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2738
Availability details for
Quantity
  • Availability
  • Add to Cart