COMH04237B66 Combi-Blocks, Inc.

2-Chloro-n-[2-(trifluoromethyl)phenyl]propanamide

  • MDL number MFCD09802007
  • Cas Number : 949444-74-0
  • Molecular Weight : 251.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2759
Availability details for
Quantity
  • Availability
  • Add to Cart