COMH04237B81 Combi-Blocks, Inc.

2-Phenyl-1,2,3,4-tetrahydroacridine-9-carboxylic acid

  • MDL number MFCD03028448
  • Cas Number : 379238-97-8
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2786
Availability details for
Quantity
  • Availability
  • Add to Cart