COMH04237BAE Combi-Blocks, Inc.

2-Amino-6-tert-butyl-n-cyclopentyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD03422651
  • Cas Number : 590356-78-8
  • Molecular Weight : 320.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2834
Availability details for
Quantity
  • Availability
  • Add to Cart