COMH04237BC8 Combi-Blocks, Inc.

2-(2-Fluorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid

  • MDL number MFCD03030478
  • Cas Number : 439140-89-3
  • Molecular Weight : 283.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2860
Availability details for
Quantity
  • Availability
  • Add to Cart