COMH04237BE7 Combi-Blocks, Inc.

5-Chloro-6-(chloroacetyl)-1,3-dimethyl-1,3-dihydro-2h-benzimidazol-2-one

  • MDL number MFCD07626505
  • Cas Number : 897545-85-6
  • Molecular Weight : 273.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2891
Availability details for
Quantity
  • Availability
  • Add to Cart