COMH04237BEF Combi-Blocks, Inc.

Methyl 2-isothiocyanato-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD03944540
  • Cas Number : 588694-54-6
  • Molecular Weight : 253.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2899
Availability details for
Quantity
  • Availability
  • Add to Cart