COMH04237C0A Combi-Blocks, Inc.

Methyl 2-(4-amino-1H-pyrazol-1-yl)acetate hydrochloride

  • MDL number MFCD12912891
  • Cas Number : 6647-89-8
  • Molecular Weight : 191.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2926

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2926
Availability details for
Quantity
  • Availability
  • Add to Cart