COMH04237C44 Combi-Blocks, Inc.

2-Amino-n-(3-nitrophenyl)thiophene-3-carboxamide

  • MDL number MFCD04055049
  • Cas Number : 669740-08-3
  • Molecular Weight : 263.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2985

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2985
Availability details for
Quantity
  • Availability
  • Add to Cart