COMH04237C47 Combi-Blocks, Inc.

5-(2,3-Dihydro-1h-indol-1-ylsulfonyl)-2-fluorobenzoic acid

  • MDL number MFCD02937103
  • Cas Number : 440343-72-6
  • Molecular Weight : 321.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2988
Availability details for
Quantity
  • Availability
  • Add to Cart