COMH04237C4F Combi-Blocks, Inc.

5-(2-Methoxyphenyl)-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD05170022
  • Cas Number : 834868-54-1
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2996
Availability details for
Quantity
  • Availability
  • Add to Cart