COMH04237C65 Combi-Blocks, Inc.

3-(2,4-Dimethylphenyl)-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD04214818
  • Cas Number : 879996-75-5
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3019
Availability details for
Quantity
  • Availability
  • Add to Cart