COMH04237C7B Combi-Blocks, Inc.

5-Ethyl-1h-1,2,4-triazole-3-thiol

  • MDL number MFCD13248675
  • Cas Number : 7271-45-6
  • Molecular Weight : 129.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3041
Availability details for
Quantity
  • Availability
  • Add to Cart