COMH04237CB9 Combi-Blocks, Inc.

4,5,6,7,8,9,10,11,12,13-Decahydrocyclododeca[b]thiophene-2-carboxylic acid

  • MDL number MFCD03945184
  • Cas Number : 588713-69-3
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3105
Availability details for
Quantity
  • Availability
  • Add to Cart