COMH04237CD2 Combi-Blocks, Inc.

(3,5-Diphenyl-1h-pyrazol-1-yl)acetic acid

  • MDL number MFCD00299897
  • Cas Number : 93323-67-2
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3130
Availability details for
Quantity
  • Availability
  • Add to Cart