COMH04237CDA Combi-Blocks, Inc.

Methyl (2-oxobenzo[cd]indol-1(2h)-yl)acetate

  • MDL number MFCD04127447
  • Cas Number : 663167-46-2
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3138
Availability details for
Quantity
  • Availability
  • Add to Cart