COMH04237CFA Combi-Blocks, Inc.

[4-(3,5-Dimethoxyphenyl)pyrrolidin-3-yl]methanol hydrochloride

  • MDL number MFCD28490093
  • Cas Number : 2173088-79-2
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3171
Availability details for
Quantity
  • Availability
  • Add to Cart