COMH04237CFB Combi-Blocks, Inc.

1-(5-Ethyl-1,3,4-thiadiazol-2-yl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD30723510
  • Cas Number : 2167328-39-2
  • Molecular Weight : 241.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3172
Availability details for
Quantity
  • Availability
  • Add to Cart