COMH04237D02 Combi-Blocks, Inc.

2-[(4-Fluorobenzyl)oxy]-3-methoxybenzoic acid

  • MDL number MFCD04488434
  • Cas Number : 861433-52-5
  • Molecular Weight : 276.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3179
Availability details for
Quantity
  • Availability
  • Add to Cart