COMH04237D08 Combi-Blocks, Inc.

3-Bromo-4-ethoxy-5-methoxybenzoic acid

  • MDL number MFCD03976236
  • Cas Number : 671790-48-0
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3185
Availability details for
Quantity
  • Availability
  • Add to Cart