COMH04237D26 Combi-Blocks, Inc.

Ethyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate

  • MDL number MFCD08142725
  • Cas Number : 864932-54-7
  • Molecular Weight : 283.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3216
Availability details for
Quantity
  • Availability
  • Add to Cart