COMH04237D49 Combi-Blocks, Inc.

2-(2-Aminoethyl)-6-(4-methoxyphenyl)pyridazin-3(2h)-one hydrochloride

  • MDL number MFCD08069899
  • Cas Number : 901886-37-1
  • Molecular Weight : 281.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3253
Availability details for
Quantity
  • Availability
  • Add to Cart