COMH04237D7C Combi-Blocks, Inc.

[5-Chloro-2-(4-propionylpiperazin-1-yl)phenyl]amine

  • MDL number MFCD06001631
  • Cas Number : 714205-78-4
  • Molecular Weight : 267.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3305
Availability details for
Quantity
  • Availability
  • Add to Cart