COMH04237DE2 Combi-Blocks, Inc.

5-(4-Butoxyphenyl)-4-ethyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD04968873
  • Cas Number : 847503-27-9
  • Molecular Weight : 277.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3407
Availability details for
Quantity
  • Availability
  • Add to Cart