COMH04237DE9 Combi-Blocks, Inc.

1-(Chloroacetyl)-4-(4-chlorophenyl)piperazine

  • MDL number MFCD01257158
  • Cas Number : 60121-78-0
  • Molecular Weight : 273.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3415
Availability details for
Quantity
  • Availability
  • Add to Cart