COMH04237E58 Combi-Blocks, Inc.

5-Chloro-1h-benzimidazole-2-sulfonic acid

  • MDL number MFCD00965565
  • Cas Number : 40828-56-6
  • Molecular Weight : 232.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3527
Availability details for
Quantity
  • Availability
  • Add to Cart