COMH04237E5D Combi-Blocks, Inc.

[(4-Butyl-2-oxo-2h-chromen-7-yl)oxy]acetic acid

  • MDL number MFCD02932376
  • Cas Number : 438027-96-4
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3532
Availability details for
Quantity
  • Availability
  • Add to Cart