COMH04237E89 Combi-Blocks, Inc.

Methyl 2-(2-methoxyphenoxy)benzoate

  • MDL number MFCD03196019
  • Cas Number : 540512-53-6
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3577
Availability details for
Quantity
  • Availability
  • Add to Cart