COMH04237ECC Combi-Blocks, Inc.

1-(4-Propoxyphenyl)pentan-1-one

  • MDL number MFCD02375363
  • Cas Number : 524732-39-6
  • Molecular Weight : 220.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3647
Availability details for
Quantity
  • Availability
  • Add to Cart