COMH04237EE5 Combi-Blocks, Inc.

N-[5-(2-Aminophenyl)-1h-1,2,4-triazol-3-yl]-4,6-dimethylpyrimidin-2-amine

  • MDL number MFCD04178129
  • Cas Number : 669719-96-4
  • Molecular Weight : 281.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3672
Availability details for
Quantity
  • Availability
  • Add to Cart