COMH04237EE6 Combi-Blocks, Inc.

N-Allyl-2-(2-chloro-4-methylbenzoyl)hydrazinecarbothioamide

  • MDL number MFCD03768434
  • Cas Number : 632292-68-3
  • Molecular Weight : 283.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3673
Availability details for
Quantity
  • Availability
  • Add to Cart