COMH04237EE7 Combi-Blocks, Inc.

N-Ethyl-4-isothiocyanatobenzenesulfonamide

  • MDL number MFCD06194931
  • Cas Number : 726144-38-3
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3674
Availability details for
Quantity
  • Availability
  • Add to Cart