COMH04237EEE Combi-Blocks, Inc.

Ethyl (2e)-(3-oxopiperazin-2-ylidene)acetate

  • MDL number MFCD01821936
  • Cas Number : 774-89-0
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3681
Availability details for
Quantity
  • Availability
  • Add to Cart