COMH04237F00 Combi-Blocks, Inc.

1-Pyridin-4-yl-piperidine-3-carboxylic acid

  • MDL number MFCD08437500
  • Cas Number : 80028-29-1
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3699
Availability details for
Quantity
  • Availability
  • Add to Cart