COMH04237F13 Combi-Blocks, Inc.

3-Methoxy-4-prop-2-ynyloxy-benzaldehyde

  • MDL number MFCD02380252
  • Cas Number : 5651-83-2
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3718
Availability details for
Quantity
  • Availability
  • Add to Cart