COMH04237F2E Combi-Blocks, Inc.

4-Methoxy-3-(phenoxymethyl)benzaldehyde

  • MDL number MFCD02055782
  • Cas Number : 438531-11-4
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3745
Availability details for
Quantity
  • Availability
  • Add to Cart