COMH04237F46 Combi-Blocks, Inc.

N-[(4-Chlorophenyl)sulfonyl]-l-leucine

  • MDL number MFCD02555823
  • Cas Number : 68305-77-1
  • Molecular Weight : 305.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3769
Availability details for
Quantity
  • Availability
  • Add to Cart