COMH04237F86 Combi-Blocks, Inc.

(5-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

  • MDL number MFCD11696367
  • Cas Number : 944897-59-0
  • Molecular Weight : 198.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3833
Availability details for
Quantity
  • Availability
  • Add to Cart