COMH04237FCE Combi-Blocks, Inc.

2-(3-Chloro-1h-1,2,4-triazol-1-yl)isonicotinic acid

  • MDL number MFCD30099286
  • Cas Number : 2090414-76-7
  • Molecular Weight : 224.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3905
Availability details for
Quantity
  • Availability
  • Add to Cart