COMH04237FDA Combi-Blocks, Inc.

2-Bromo-n-(2,4-dimethoxyphenyl)acetamide

  • MDL number MFCD02676798
  • Cas Number : 349120-93-0
  • Molecular Weight : 274.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3917
Availability details for
Quantity
  • Availability
  • Add to Cart