COMH04237FDE Combi-Blocks, Inc.

5-(4,5-Dimethylthien-3-yl)-4-isopropyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD03423435
  • Cas Number : 847503-20-2
  • Molecular Weight : 253.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3921
Availability details for
Quantity
  • Availability
  • Add to Cart