COMH04237FEE Combi-Blocks, Inc.

4-Methyl-2-phenyl-5-(pyridin-3-ylmethyl)-1h-pyrazolo[4,3-c]pyridine-3,6(2h,5h)-dione

  • MDL number MFCD06755313
  • Cas Number : 847572-55-8
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3937
Availability details for
Quantity
  • Availability
  • Add to Cart