COMH04237FFB Combi-Blocks, Inc.

4,4,4-Trifluorobutanoyl chloride

  • MDL number MFCD00092810
  • Cas Number : 406-91-7
  • Molecular Weight : 160.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3950
Availability details for
Quantity
  • Availability
  • Add to Cart