COMH04238034 Combi-Blocks, Inc.

3-Hexyl-7-hydroxy-4,8-dimethyl-2h-chromen-2-one

  • MDL number MFCD02331303
  • Cas Number : 405918-92-5
  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4007
Availability details for
Quantity
  • Availability
  • Add to Cart